Learn More
Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems
For visible bulk drying
Description
Description
For visible bulk drying
  • Durable and corrosion-resistant
  • Chambers without valves include clear acrylic top and bottom lids, gaskets, vacuum break valve, and electrical pass-through for transformer cords
  • Chambers with valves include aluminum lid with eight valves that accommodate both 1/2 in. and 3/4 in. adapters for connecting flasks
  • Use with or without product shelves (shelves available separately, visit www.fishersci.com)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems