missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Drying Train: Tubing Enclosure Kit
Accessories for Labconco Drying Train
Brand: Labconco™ 5242501
This item is not returnable.
View return policy
Description
For all glove boxes
Composed of 25 ft. fluorinated ethylene propylene (FEP) tubing and eight inserts
Specifications
Labconco Drying Train | |
3 lb. | |
25 ft. FEP tubing, eight tube inserts and two connectors to connect the oxygen and moisture removal column to the diaphragm pump |
All glove box | |
1.4 kg | |
Tubing Enclosure Kit |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labconco™ Drying Train: Tubing Enclosure Kit > Oxygen and moisture drying train tubing Kit
Spot an opportunity for improvement?Share a Content Correction