Learn More
Labconco™ Ducting Parts: Connectors
Connectors for Labconco ducts
Brand:  Labconco™ 5440900
605.40 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 15789688
  • £899.00 / Each
Explore more special offers
Description
Description
Powder-coated steel assembly for connecting a glove box to a building's exhaust or a remote blower. 6 in. diameter connection.
Specifications
Specifications
Hoods and Enclosures
Connector, Exhaust (Six Thimble)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Ducting Parts: Connectors > Connector, Exhaust (six thimble)