Learn More
Labconco™ Flat Work Surface
Extend your work space with the Labconco Flat Work Surface, perfect when paired with the Labconco Fiberglass 30 and Basic™ hoods
Brand:  Labconco™ 4882804
 View more versions of this product
Product Code. 16647142
  • £3100.00 / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Extend your work space with the Labconco Flat Work Surface, perfect when paired with the Labconco Fiberglass 30 and Basic™ hoods
Install atop Labconco Protector™ storage cabinets and adjustable height base stands.
  • Placed atop two Protector standard base cabinets to support precise glove boxes
  • Flat, solid-epoxy work surfaces
Specifications
Specifications
Flat Work Surface
Without Cutout
76 cm
1.25 in.
30 in.
93 kg
183 cm
Black
30 in.
Basic 70 laboratory hood, Precise™ and XPert™ weigh box
3 cm
205 lb.
72 in.
Epoxy