Learn More
Labconco™ Freeze-Drying Ampule Accessories
Product Code. p-21252 Shop All Labconco Corporation Products
Click to view available options
Type:
Sealing Torch
Unit Size:
Each
Product Code. 15123075
Brand: Labconco™ 7578500
Only null left
Add to basket
Add to basket
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
Type Sealing Torch
Material Stainless Steel
For Use With (Application) Connects to natural gas, butane, or propane and oxygen
For Use With (Equipment) Freeze drying system
Weight (English) 3 lb.
Weight (Metric) 1.4 kg