Learn More
Labconco™ HEPA Filter
For Purifier Class I and HEPA-Filtered Enclosures and XPert and RXPert Enclosures.
Brand: Labconco™ 3707902
Code : NEW
Additional Details : Weight : 71.70000kg
Description
Supply HEPA filter for 4' Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert & RXPert Filtered Balance Systems.Specifications
99.99% | |
4 ft. Purifier™ class I safety, HEPA filtered, vertical clean bench and PCR enclosure | |
71.7 Kg | |
101.6 to 127 cm | |
42 in. | |
3.3 in. |
HEPA | |
158 lb. | |
40 to 50 in. | |
18 in. | |
Particulate |