Learn More
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly with Standard Taper 19/38 Joints
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring.
£349.00 - £546.00
Specifications
Material Borosilicate Glass
Includes Glass top and bottom, Silicone Rubber ring seal
Product Type Flask Kit
Standard Taper Joint 19/38
Weight (English) 3 lb.
View More Specs
Products 7
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15103035
View Documents
Labconco™
7550000
25 mL
£349.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15113035
View Documents
Labconco™
7550200
50 mL
£356.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15123035
View Documents
Labconco™
7550400
100 mL
£364.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15153035
View Documents
Labconco™
7550900
750 mL
£504.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15163035
View Documents
Labconco™
7551000
1000 mL
£546.00
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
15133035
View Documents
Labconco™
7550600
250 mL
£392.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15143035
View Documents
Labconco™
7550800
500 mL
£504.00
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Description
Description
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring
  • Flasks should be filled no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is assured.
  • Adapters are required to attach flasks to freeze dry valve ports (sold separately).
Specifications
Specifications
Borosilicate Glass
Flask Kit
3 lb.
Joint
1.4 kg
Glass top and bottom, Silicone Rubber ring seal
19/38
Reusable
Flat
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly with Standard Taper 19/38 Joints