Learn More
Labconco™ Manual Hydraulic Lift Base Stands
Manual Hydraulic Lift Base Stands for Labconco Purifier and XPert Enclosures.
Description
Description
These stands have a built-in hydraulic lift so that height may be adjusted at any time. They allow you to position the stand and the safety enclosure or clean bench it supports to accommodate persons of different heights and wheelchair-bound users, to transport equipment through doorways, and for easier service access. • Support Purifier Class I Safety Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, Purifier Filtered PCR Enclosures, XPert Balance Enclosures, XPert Nano Enclosures, and XPert and RXPert Filtered Balance Systems. • Support loads up to 1000 pounds. • ADA-compliant. • Durable 1.75 in. tubular, powder-coated steel frame with rear crossbar supports. Sub-frame is 2.0 in. x 3.0 in.. • Height is infinitely adjustable from 25.5 in. to 33.5 in. high. • Include four powder-coated, die-cast feet with levelers. • Include front-accessible hand crank. Turning the crank clockwise raises the stand; counterclockwise lowers it. The handle may be disengaged and folded under, out of the way, when not in use.
1Use only with flat work surface.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Labconco™ Manual Hydraulic Lift Base Stands