missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ PrimeMate™ Vacuum Pump Oil

For use with the Labconco PrimeMate Oil Change System

£415.00 - £566.00

Specifications

For Use With (Equipment) PrimeMate Oil Change System
Volume (Metric) 3.8 L
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
17104534
View Documents
Labconco™
1988001
Vacuum Pump Oil
£415.00
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
15103025
View Documents
Labconco™
7541300
Synthetic Vacuum Pump Oil
£566.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Choice of one gallon (3.8L) hydro-treated paraffinic hydrocarbon oil or synthetic hydrocarbon oil
  • Recommended for Labconco general purpose vacuum pumps

Specifications

Specifications

PrimeMate Oil Change System
3.8 L
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ PrimeMate™ Vacuum Pump Oil

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.