Learn More
Labconco™ Purifier™ Vertical Clean Bench
Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.
£7595.00 - £11105.00
Specifications
Product Type | Clean Bench |
---|---|
Coating | Powder |
Filter Type | HEPA |
Lighting Type | Fluorescent |
Material | Powder-coated Aluminum and Steel |
Alert:
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
Requires: Work surface and base stand (sold separately).
Product Code | Brand | Width (English) | Voltage | Plug Type | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (English) | Voltage | Plug Type | Includes | Price | Quantity & Availability | ||||||
17134871
|
Labconco™
3970220 |
24 in. | 230 V | No Plug | Bench Only | N/A | N/A | N/A | |||||
17144871
|
Labconco™
3970320 |
36 in. | 230 V | No Plug | Bench Only | N/A | N/A | N/A | |||||
15764555
|
Labconco™
3970421 |
48 in. | 230 V | No Plug | UV light, protection panel and timer | N/A | N/A | N/A | |||||
Description
- Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
- Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
- ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
- Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
- Intrinsically-sound, negative pressure design
- Minihelic pressure gauge and disposable prefilter
- UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
- UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
- Light and blower switches
- Air diffuser and 23.8cm (9.4 in.) high sash opening
- Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
- Two utility ports with iris openings and plugs
- ETL Listing (115 volt, 60 Hz models)
Order Info
Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.
Warranty
One year parts and service
Specifications
Clean Bench | |
HEPA | |
Powder-coated Aluminum and Steel | |
Glacier White, Gray | |
25.3 in. | |
64.3 cm | |
36.8 in. | |
93.47 cm | |
50/60 Hz | |
Utility |
Powder | |
Fluorescent | |
CAN/CSA C22.2, UL, CE | |
28.6 in. | |
72.64 cm | |
Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation | |
22.7 in. | |
57.7 cm | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.