Learn More
Labconco™ Replacement Bag for Hepa Filter Cell
Replacement bag for filters in XPert and RXPert Balance and Bulk Powder Enclosures and XPert Nano Enclosures.
Brand:  Labconco™ 3776002
Additional Details : Weight : 0.10000kg
Product Code. 10027942
  • £388.00 / Each
Estimated Shipment: 28-10-2024
to see stock.
Description
Description
Replacement Bag for Bag-in/Bag-out HEPA Filter for use with Labconco FilterMate Portable Exhausters.
Specifications
Specifications
HEPA
1 lb.
Filter Accessory
HEPA filter cell, Protector™ demonstration hood, FilterMate™ portable exhauster
0.5 Kg
Bag-in/bag-out HEPA filter to contain particulate matter during filter changing