Learn More
Labconco™ Sash Stop Kit
A Sash Stop restricts how far a vertical-rising sash may be opened.
Brand:  Labconco™ 9410300
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 17601037
  • £25.84 / Each
Estimated Shipment: 30-09-2024
to see stock.
Description
Description
Sash stop kit for use on most Labconco fume hoods made since 2012. 1 kit required for 3' to 8' Premier, XL Benchtop, XStream, PVC, Stainless Steel and XL Floor-Mounted Fume Hoods with vertical-rising sashes. 2 kits required for 10', 12' XL Benchtop with dual sash, Pass-Through Fume Hoods. A Sash Stop restricts how far a vertical-rising sash may be opened. This small plastic device may be easily field installed on the fixture panel of a Protector Hood, which have been factory-prepared to accommodate a sash stop at the 60% open position (18 in. to work surface). Alternate sash positions may be field drilled. Each Sash Stop Kit includes components for one sash.
Specifications
Specifications
Protector™ Premier™, Protector™ XStream™ fume hood
6 in.
30.48 cm
15.24 cm
Plastic
12 in.
6 in.
6 x 6 x 12 in. (15.24 x 15.24 x 30.48 cm)
15.24 cm