Learn More
Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream
SpillStopper work surfaces are dished to contain spills
Brand:  Labconco™ 9503400
 View more versions of this product
Product Code. 15963117
  • £2070.00 / Each
Explore more special offers
Description
Description
  • Front edge has large radius to aerodynamically direct airflow in the hood
  • Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
Specifications
Specifications
No Cutouts
Epoxy Resin