missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Label Maker Cartridge: Nylon Cloth
Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
£28.65 - £87.30
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15161899
|
Brady™
143361 |
0.5 x 1.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12823125
|
Brady™
143350 |
1 x 0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11992852
|
Brady™
143355 |
0.5 x 1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12789536
|
Brady™
143351 |
12.7 mm x 4.9 m (W x L) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15152907
|
Brady™
143360 |
19.05 mm x 4.9 m (W x L) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12754049
|
Brady™
143352 |
0.75 x 0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12734059
|
Brady™
143362 |
1.25 x 1.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15172907
|
Brady™
143357 |
25.4 mm x 4.88 m (W x L) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11942852
|
Brady™
143348 |
0.825 x 0.375 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12719546
|
Brady™
143356 |
1 x 0.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11982852
|
Brady™
143354 |
1 x 0.375 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15132907
|
Brady™
143349 |
9.53 mm x 4.9 m (Wx L) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Label Maker Cartridge: Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
- Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
- Recommended for indoor use
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- All-in-one cartridge includes ribbon and label supply
Specifications
Nylon Cloth |