missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Labmaster Air-Drive Mixer
Use Labmaster Air-Drive Mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical. These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.
£1655.00 - £1825.00
Specifications
Speed | 1800 rpm |
---|---|
Horsepower | 1/15 hp |
Material | 316 Stainless Steel |
Motor Type | Air-Driven |
Torque | 37 in. oz. |
Product Code | Brand | Mounting Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mounting Type | Price | Quantity & Availability | |||||
15457463
|
Cole-Parmer™
50322-00 |
Tube Suspension |
£1655.00
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15467463
|
Cole-Parmer™
50322-10 |
Universal Clamp |
£1825.00
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
- These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes.
- LABMASTER air-drive mixers have two types of mounting styles.
- The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately).
- The universal clamp mixers mount directly to a ring stand or tank rim.
- Mixers include:
- 5/16 x 12 in., 316 SS nonthrough-type shaft
- 5/16 x 2.5 in., 316 SS A-310 impeller
- Collet-type chuck
- Universal clamp or tube suspension mounting
- Muffler
- Needle value
- Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
1800 rpm | |
316 Stainless Steel | |
37 in. oz. |
1/15 hp | |
Air-Driven | |
Mixer |