missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Lab Washer PG 8583 CD AE WW APD CM
Optimally designed laboratory washer with perfectly aligned wash pressure and ideally designed baskets, filters, and spray arms. Miele™ PG 8583 CD AE WW APD CM Laboratory Glassware Washer features a variety of specific wash programs to meet multiple laboratory needs.
Brand: Miele Professional™ 62858361
14563.59 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Variable-speed heater pump
- Efficient hot air drying Dry Plus
- Optimum cleaning performance
- Optimum wash pressure
- Low consumption values
- Short batch times
- Crevice-free wash cabinet
- Spray arm pressure monitoring
- Spray arm monitoring
- Hot water connection
- De-min. water connection
- Liquid dispensing
- Conductivity monitoring
Specifications
Freestanding or built-under | |
500 L/min. | |
600 mm | |
820 mm | |
3 AC 208V 60HZ |
Stainless Steel | |
IP safety class according to EN 60529: IP 21, EN 61010-1, EN 61010-2-040, RoHS Directive 2011/65/EU, c CSA us | |
474/520 x 530 x 520mm | |
900 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction