missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Lab Washer PG 8593 WW AD

Optimally designed laboratory washer with perfectly aligned wash pressure and ideally designed baskets, filters, and spray arms. Miele™ PG 8593 WW AD Laboratory Glassware Washer features a variety of specific wash programs to meet multiple laboratory needs.

Brand:  Miele Professional™ 62859301

Product Code. 15477694

  • £11620.00 / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Optimally designed laboratory washer with perfectly aligned wash pressure and ideally designed baskets, filters, and spray arms. Miele™ PG 8593 WW AD Laboratory Glassware Washer features a variety of specific wash programs to meet multiple laboratory needs.

  • Variable-speed heater pump
  • DryPlus
  • Optimum cleaning performance
  • Optimum wash pressure
  • Low consumption values
  • Short batch times
  • Crevice-free wash cabinet
  • Spray arm pressure monitoring
  • Spray arm monitoring
  • Liquid dispensing
Specifications

Specifications

Freestanding or built-under
Stainless Steel
IP safety class according to EN 60529: IP 21, EN 61010-1, EN 61010-2-040, RoHS Directive 2011/65/EU, c CSA us
70°C
60.0 cm
835 mm
45 A
400 V
Digital
500 L/min.
Labware/reusable glassware washing
23.62 in.
474/520 x 530 x 520 mm
600 mm
400 V, 3 Phase, 45 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers