Learn More
Smeg™ Laboratory Glassware Washer
Clean a wide range of glassware with this washer. Smeg™ Laboratory Glassware Washers are manufactured with high quality materials and feature faultless, intuitive performance. Specific accessories ensure high quality washing of even the most difficult and dirtier instruments.
POA
Specifications
Material | Stainless Steel |
---|---|
Product Type | Glassware Washer |
Dimensions (D x W x H) Interior | 490 x 525 x 570mm |
Description
- Microprocessor electronic control
- Active thermodynamic drying
- Alkaline powder detergent dispenser
- Peristaltic pump for acid liquid neutralizer
- Washing chamber and internal door in AISI 304 stainless steel
- External panels made in AISI 304 stainless steel
- 2 independent washing levels through telescopic guides
- Incorporated water softener with salt sensor
- Cold water and demineralized water connections
- Electronic door lock system for user safety
- Without trolleys
Specifications
Stainless Steel | |
Glassware Washer | |
490 x 525 x 570mm |