Learn More
Fisherbrand™ Laboratory Jacks
Provides for precise height adjustment with the turn of a knob. Fisherbrand™ Laboratory Jacks are manufactured from aluminum alloy with a lifting screw in stainless steel.
Brand:  Fisherbrand™ 2046FISH
 View more versions of this product
Product Code. 11517023
  • £455.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Plastic knob operates the scissor action lifting mechanism
  • Either plate can be used as the base
  • Available in a range of plate dimensions
Specifications
Specifications
Aluminum
280 mm
35 kg
Laboratory Lift
1 Ea.
60 to 450 mm (Top Plate)
260 mm
Stainless steel Lifting screw
12mm dia. Support Rod