Learn More
Cytiva (formerly Pall Lab) Laboratory Manifold Coupling Device
Accessory coupling device for microbiology manifold.
Brand:  Cytiva (formerly Pall Lab) 4893
Additional Details : Weight : 0.19950kg
Product Code. 17144441
  • £332.00 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket
Description
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifications
Specifications
Stainless Steel (316L)
Non-sterile