missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 19cm Diamter

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 19cm Diameter offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

POA

Specifications

Dimensions (L x W) 19 x 19 cm
Type Lab Safety Mat
Quantity 50/Pk.
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15707079
View Documents Promotion Details
Dycem™
LS02194
Silver
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15717079
View Documents Promotion Details
Dycem™
LS02196
Black
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15727079
View Documents Promotion Details
Dycem™
LS02198
Yellow
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15787069
View Documents Promotion Details
Dycem™
LS02191
Blue
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15797069
View Documents Promotion Details
Dycem™
LS02192
Red
POA
Pack of 50
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Anti-slip properties
  • 19cm diameter
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors
Specifications

Specifications

19 x 19 cm
Lab Safety Mat
50/Pk.
Videos
SDS
Documents

Documents

Product Certifications