missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 35 x 25cm

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 35 x 25cm offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

POA

Specifications

Dimensions (L x W) 35 x 25 cm
Type Lab Safety Mat
Quantity 10/Pk.
View More Specs

Products 4
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15707089
View Documents Promotion Details
Dycem™
NS02MD8
Yellow
POA
Pack of 10
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15777079
View Documents Promotion Details
Dycem™
NS02MD1
Blue
POA
Pack of 10
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15787079
View Documents Promotion Details
Dycem™
NS02MD2
Red
POA
Pack of 10
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15797079
View Documents Promotion Details
Dycem™
NS02MD6
Black
POA
Pack of 10
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Anti-slip properties
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors
Specifications

Specifications

35 x 25 cm
Lab Safety Mat
10/Pk.
Videos
SDS
Documents

Documents

Product Certifications