Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Clear Body, Tear-Off Protection Strip
Designed to collect samples to detect the presence of microbiological contaminations
£214.08
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17976469
|
Labplas
KSS-67110-DE |
DE Neutralizing buffer |
£214.08
Pack of 100 |
Estimated Shipment: 10-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Detects Listeria, Salmonella and E. coli
- Contaminant-free
- Virgin polyethylene
- Large bag mouth opening
- Easy to use pull tabs
- Sponge for swabbing
- Dehydrated sponge
Specifications
450 mL | |
Polyethylene |