missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labplas Twirl'EM™ Sterile Sampling Bags, SafetyTabs
Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM™ Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.
Brand: Labplas EPL47012
This item is not returnable.
View return policy
Description
- Virgin polyethylene tubing
- Can be heated to 240°C for sterility
- Large bag mouth opening to facilite insertion of samples
- Easy pull tabs to open and close the bag with decreased risk of contamination
- R-Nase, D-Nase, Pyrogen, and BPA free
- Environmental sampling
- Surface sampling
- Carcass sampling
- Biomedical research
- Pharmaceutical research
- Food industry
Certifications
Sterility
Compliance
EPA, FDA, USDA, HACCP
Specifications
Clear body | |
2 Round Wires | |
Sterile |
1650 mL | |
Polyethylene | |
4 mil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction