missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, 3 ports, Single Pack
GREENER_CHOICE

Product Code. p-200003876 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
2 L
5 L
10 L
20 L
50 L
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17819746

Brand: Thermo Scientific™ SH31295.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific 2D Labtainer™ with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:

  • Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
  • Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
  • Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:

  • Harvest from, feed into, and collect samples from bioreactors and fermentors
  • Buffer, culture media, and process liquid preparation and storage
  • Chromatography feed and fraction collection
  • Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology

Choose from the industry's largest component library with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters
  • Tubing—thermoplastic elastomers, platinum-cured silicone
  • Sample ports—septum-style and needle-free ports
  • Clamps—sliders, pinch, and crimp bands

Line 1

MPC insert, polycarbonate

Tubing length: 30 cm (12 in.)

ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)

Line 2

MPC insert, polycarbonate

Tubing length: 30 cm (12 in.)

ID x OD: 9.7 x 12.7 mm (0.38 x 0.5 in.)

Line 3

End plug, polypropylene

Tubing length: 8 cm (3 in.)

ID x OD: 6.35 x 10.92 mm (0.25 x 0.43 in.)

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Quantity 2 L
Tubing Length Line 1 30 cm (12 in.)
Tubing Length Line 3 8 cm (3 in.)
Tubing Length Line 2 30 cm (12 in.)
Tubing Material Line 3 Polypropylene
Tubing Material Line 1 Polycarbonate
Tubing Material Line 2 Polycarbonate
Tubing Style TPE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.