missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container, BioTitan retainers, two ports, Luer locks

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Brand:  Thermo Scientific™ SH31292.15

 View more versions of this product

Product Code. 17839686

This item is not returnable. View return policy

Description

Description

The Thermo Scientific 2D Labtainer with BioTitan BioProcess Container is composed of Aegis5-14 single-web, multi-layer film.

Features of the Thermo Scientific 2D BPC system include:
• Now manufactured with the BioTitan Retention Device—designed to provide superior universal retention that minimizes the risk of leakage and failure at the connection point
• Use to eliminate post-use cleaning steps required with re-useable containers and to reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Thermo Scientific 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Buffer, culture media, and process liquid preparation and storage
• Chromatography feed and fraction collection
• Harvest, storage, and transport of bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 L
TPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers