Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, three ports, Luer locks
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
£211.77 - £1123.05
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | ISO 7 |
Color | Transparent |
Disposable | Single-use |
For Use With (Application) | Bioprocessing |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17870607
|
Thermo Scientific™
SH31050.11 |
50 mL |
£372.45
Pack of 10 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17880667
|
Thermo Scientific™
SH31050.12 |
100 mL |
£211.77
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800677
|
Thermo Scientific™
SH31050.13 |
250 mL |
£224.96
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890607
|
Thermo Scientific™
SH31050.14 |
500 mL |
£295.90
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800617
|
Thermo Scientific™
SH31050.15 |
1 L |
£314.45
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820677
|
Thermo Scientific™
SH31050.16 |
2 L |
£324.22
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840607
|
Thermo Scientific™
SH31049.01 |
5 L |
£684.11
Pack of 10 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17870667
|
Thermo Scientific™
SH31049.02 |
10 L |
£694.09
Pack of 10 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17880607
|
Thermo Scientific™
SH31049.03 |
20 L |
£659.18
Pack of 10 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860667
|
Thermo Scientific™
SH31049.04 |
50 L |
£1123.05
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890667
|
Thermo Scientific™
SH31049.05 |
2 L |
£529.78
Pack of 10 |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Not Autoclavable | |
Transparent | |
Bioprocessing | |
C-Flex | |
3 ports |
ISO 7 | |
Single-use | |
Sterile | |
Aegis 5-14 |