Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock insert and body connectors, injection site
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Brand: Thermo Scientific™ SS00003-I
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors

Specifications
Bioprocess Container | |
500 mL | |
EVA | |
ASI™ 28 | |
Luer lock connectors and injection site | |
50/Cs. | |
Irradiated | |
8 in. | |
6.5 in. |
Store at room temperature. | |
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
EVA | |
One 4-mil. poly bag | |
Labtainer™ | |
Room Temperature | |
20.3 x 16.5 cm (8.0 x 6.5 in.) | |
20.3 cm | |
16.5 cm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.