missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00010-I

 View more versions of this product

Product Code. 30067382

  • £1640.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bioprocess Container
2 L
EVA
ASI™ 28
Luer lock connectors, reducer and injection site
50/Cs.
Irradiated
33.3 cm
20.3 cm
Store at room temperature.
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
EVA
One 4-mil. poly bag
Labtainer™
Room Temperature
33.3 x 20.3 cm (13.1 x 8.0 in.)
8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers