missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors, injection site
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
£20.35 - £1385.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Material | EVA |
Tubing Style | EVA |
Film Description | ASI™ 28 |
Outer Packaging | One 4-mil. poly bag |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17820787
|
Thermo Scientific™
SS00011-I |
100 mL | N/A | ||||||
17800787
|
Thermo Scientific™
SS00014-I |
1 L |
£1385.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Store at room temperature. | |
EVA | |
One 4-mil. poly bag | |
Labtainer™ | |
Irradiated |
EVA | |
ASI™ 28 | |
MPC insert and body connectors and injection site | |
Room Temperature |