missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00018-I

Additional Details : Weight : 4.20000kg

Product Code. 15949906

  • POA / 25 units
Estimated Shipment: 15-10-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications

Specifications

Store at room temperature.
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
EVA
One 4-mil. poly bag
Labtainer™
Room Temperature
63.3 x 42.7 cm (24.9 x 16.8 in.)
16.8 in.
20 L
Ethyl Vinyl Acetate
ASI™ 28
MPC insert and body connectors, plug and injection site
25/Cs.
Irradiated
63.3 cm
42.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers