missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00016-I

Product Code. 15929906

  • POA / 30 units

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
EVA
One 4-mil. poly bag
Labtainer™
Room Temperature
31.5 x 32.0 cm (12.4 x 12.6 in.)
12.6 in.
5 L
Ethyl Vinyl Acetate
ASI™ 28
MPC insert and body connectors, plug and injection site
30/Cs.
Irradiated
31.5 cm
32 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site > 5L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.