Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100mL with 2 Ports, Luer Lock, 2D BPC

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.
POA
Specifications
| Dimensions | 14.7 cm x 14.2 cm (5.8 in. x 5.6 in.) |
|---|---|
| Film Description | CX5-14 |
| Port Detail | Edge Ports: 2 x 0.25 in. |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Product Line | Labtainer™ |
| Product Code | Brand | Capacity (Metric) | Tubing Style | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Tubing Style | Price | Quantity & Availability | ||||
11506311
![]() |
Thermo Scientific™
SH30658.12 |
100 mL | 12 in. of 1/4 in. Tubing |
POA
Pack of 10 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
11546301
![]() |
Thermo Scientific™
SH30657.12 |
100 mL | No Tubing |
POA
Pack of 10 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock and MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers and process intermediates
Separation – feeding and receiving of sterile liquids from separation systems
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
| 14.7 cm x 14.2 cm (5.8 in. x 5.6 in.) | |
| Edge Ports: 2 x 0.25 in. | |
| Labtainer™ | |
| 10 per case | |
| 100 mL |
| CX5-14 | |
| Packages placed in strong cardboard cartons with product and lot identification | |
| Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
| Gamma Irradiated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.