Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, Luer Lock, 2D BPC
Simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.
Brand: Thermo Scientific™ SH30713.02
545.04 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Using Thermo Scientific™ Labtainer™ BPC bag with 3 ports (luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.
Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers and process intermediates
Separation – feeding and receiving of sterile liquids from separation systems
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
62.2 cm x 30 cm (24.5 in. x 11.8 in.) | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
Gamma Irradiated | |
Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.) | |
Labtainer™ | |
Edge Ports: 2 x 0.38 in. 1 x 0.25 in. |
Packages placed in strong cardboard cartons with product and lot identification | |
1 | |
Line 1. Insert luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.) | |
Line 2. Body luer lock connection, polypropylene Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.) | |
Fewer resources used and less waste | |
CX5-14 |