missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC

Using these BPC bags with 3 ports (MPC insert, MPC body, luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Brand:  Thermo Scientific™ SH30712.02

Product Code. 17895736

  • £725.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

10 L
Packages placed in strong cardboard cartons with product and lot identification
Gamma Irradiated
Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 3.2 x 6.4 mm  (0.13 x 0.25 in.)
Labtainer™
Edge Ports: 2 x 0.38 in. 1 x 0.25 in.
62.2 cm x 30 cm (24.5 in. x 11.8 in.)
10 L, each
Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
Fewer resources used and less waste
CX5-14
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, MPC Insert, MPC Body, and Luer Lock, 2D BPC > Size: 10L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.