missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 1L with 2 Ports, Luer Lock and MPC Insert, 2D BPC

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Brand:  Thermo Scientific™ SH30662.15

Product Code. 17885706


Description

Description

Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers, and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

TRUSTED_SUSTAINABILITY
Specifications

Specifications

29.7 cm x 20.1 cm (11.7 in. x 7.9 in.)
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Gamma Irradiated
Line 2. MPC insert, polycarbonate, No Tubing
Edge Ports: 2 x 0.25 in.
Packages placed in strong cardboard cartons with product and lot identification
10 per case
Line 1. Body luer lock connection,polypropylene Tubing: C-Flex, length: 30 cm (12 in.) Id x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Labtainer™
CX5-14
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers