Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 2 ports, capsule filter and MPC insert
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Brand: Thermo Scientific™ SS00162-I
Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 membrane capsule filter and 1 MPC insert with pinch clamps). It is available in 50, 100 and 200L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: C-Flex 374, length: 91.4cm (36 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC insert + cap, pinch clamp. Tubing: C-Flex 374, length: 91.
Specifications
| Store at room temperature. | |
| 2 Ports (1 Membrane Capsule Filter and 1 MPC Insert With Pinch Clamps) | |
| C-Flex, 91.4 cm (36 in.) | |
| Two 4-mil. poly bags | |
| Labtainer™ | |
| 5/Cs. | |
| Irradiated | |
| 37.5 in. | |
| 21.5 in. |
| 50 L | |
| Polyethylene | |
| ASI™ 26/77 | |
| Capsule filter and MPC insert + cap | |
| Protective material around rough areas | |
| Room Temperature | |
| 95.3 x 54.6 cm (37.5 x 21.5 in.) | |
| 95.3 cm | |
| 54.6 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.