Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Brand: Thermo Scientific™ SS00040-I
Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 MPC insert + cap, 1 MPC body + cap, 1 injection site). Available in 10L or 20L capacity.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: MPC insert with 0.97cm (0.38 in.) barb, cap, and pinch clamp. Tubing: TPE 374, length: 30cm (12 in.), ID x OD: 9.5 x 16mm (0.38 x 0.63 in.)
- Line 2: MPC body with 0.97cm (0.38 in.) barb, plug, and pinch c
Specifications
Store at room temperature. | |
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
TPE, 30 cm (12 in.) | |
One 4-mil. poly bag | |
Labtainer™ | |
Room Temperature | |
32 x 56.5 cm (12.6 x 22.3 in.) | |
22.3 in. |
10 L | |
Polyethylene | |
ASI™ 26/77 | |
MPC connections and injection port | |
25/Cs. | |
Irradiated | |
32 cm | |
56.5 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.