Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
POA
Specifications
Content And Storage | Store at room temperature. |
---|---|
Material | Polyethylene |
Tubing Style | C-Flex, 51 cm (20 in.) |
Film Description | ASI™ 26/77 |
Outer Packaging | 4 mil. poly bag, One 4-mil. poly bag |
Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap). It is available in 50, 100 and 200L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: MPC insert with 9.5mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51cm (20 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC body with 9.5mm (3/8 in.) barb, plug, and pinch clamp.
Specifications
Store at room temperature. | |
C-Flex, 51 cm (20 in.) | |
4 mil. poly bag, One 4-mil. poly bag | |
Labtainer™ | |
Room Temperature |
Polyethylene | |
ASI™ 26/77 | |
MPC connections + caps, pinch clamps | |
Poly bags on tubing ends, protective material around rough areas | |
Irradiated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.