missing translation for 'onlineSavingsMsg'
Learn More

LACTB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen LACTB2
Concentration 0.29 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16836524
View Documents
Proteintech
16783-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16826524
View Documents
Proteintech
16783-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.
Specifications

Specifications

LACTB2
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Rat, Human
Q53H82, Q561R9, Q99KR3
212442, 297768, 51110
LACTB2 Fusion Protein Ag10213
Primary
-20°C
LACTB2
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Beta lactamase like protein 2, CGI 83, lactamase, beta 2, LACTB2
LACTB2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications