missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, 85%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
232.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Formula Weight | 90.08 |
Percent Purity | 85% |
Grade | BAKER ANALYZED™ ACS Reagent |
Quantity | 500 mL |
Flash Point | 112.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.20kg |
Chemical Name or Material | Lactic Acid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'