Learn More
3M™ Petrifilm™ Lactic Acid Bacteria Count Plate
A time-saving, sample-ready plate designed to determine total lactic acid bacteria populations in food and environmental samples.
Brand:  3M™ Petrifilm™ 7100088605
Encompass_Preferred
 View more versions of this product
Product Code. 17243774
  • £120.76 / Pack of 50
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Results in 48 hours of incubation time.
  • Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes.
  • Self-contained anaerobic environment enabled by oxygen-scavenging technology and oxygen-barrier films. No gas packs, chambers, or CO2 incubators needed.
  • This plate has been awarded Performance Tested Method (PTM) #041701 for selected foods and NF VALIDATION 3M 01/19-11/17 by AFNOR Certification for all human foods (excluding yogurts) and industrial environmental samples.
  • All-in-one method for differentiation of gas-producing colonies without Durham tubes or extra steps.
  • Proven reliability versus MRS and APT agar methods. Can be used to test a broad range of foods and environmental samples.
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Environmental Test
50