missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
87.15 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Description
Lactic Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 1,2 |
Boiling Point | 125°C |
Color | Yellow |
Formula Weight | 90.08g/mol |
Quantity | 500 mL |
Packaging | HDPE plastic bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'