missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, Honeywell Fluka™
BIOPHARMA

Tested according to Ph.Eur.

£61.09 - £150.92

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15636730
BIOPHARMA
View Documents
Honeywell-Fluka
69775-250ML
250 mL
£61.09
250mL
Please to purchase this item. Need a web account? Register with us today!
15626730
BIOPHARMA
View Documents
Honeywell-Fluka
69775-1L
1 L
£150.92
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

50-21-5
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
612
2-hydroxypropanoic acid
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CHEBI:78320
CC(O)C(O)=O

Specifications

50-21-5
CH3CH(OH)COOH
NONH for all modes of transport
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CC(O)C(O)=O
90.08
CHEBI:78320
1.209g/mL at 25°C
Lactic acid
C3H6O3
MFCD00004520
1209341
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
612
90.08g/mol
113°C (235.4°F)
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338

Product Identifier
  • Lactic acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Lactic acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.