Learn More
Himalayan™ Ladies Black Star Safety Boot with Midsole
Black safety boot with toecap protection and slip resistant outsole.
Brand:  Himalayan™ 2211/5
Encompass_Preferred
 View more versions of this product
Product Code. 15407560
  • POA / 1 pair
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • 200 Joule toecap protection
  • SRC - outsole slip resistance
  • Steel midsole
  • Oil resistant
  • Heat resistant to 200°C
  • EN ISO 20345-S1P
Specifications
Specifications
Black
5