missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Laemmli SDS sample buffer, reducing (6X)

Product Code. 15403949
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
This item is not returnable. View return policy

Product Code. 15403949

Brand: Thermo Scientific Chemicals J61337.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.

For protein sample preparation to be used in the Laemmli SDS-PAGE system. Laemmli SDS sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Further, it is used for polyacrylamide protein gel analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • This compound is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Concentration 6X
Product Type Sample Loading Buffer
Form Liquid
For Use With (Application) Laemmli SDS-PAGE
Quantity 50 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Laemmli SDS sample buffer, reducing (6X)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate, 2-Mercaptoethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.