Learn More
LAG-3 (CD223) Mouse anti-Human, PE-CF594, Clone: T47-530, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 565719
Product Code. 17267484
  • ONLINE EXCLUSIVE PRICE
    £141.00
    £134.83 / 25 tests
    Save £6.17
    4% Off
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAG-3 (CD223)
Monoclonal
PE-CF594
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC
Human LAG-3 Recombinant Protein
RUO
Human
IgG1 κ
Flow Cytometry
T47-530
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD HorizonTM PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.