Learn More
Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor
Plate filling from reagent reservoirs
Brand:  Corning™ 6070
Product Code. 17842151
  • £25185.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor can perform a variety of liquid handling tasks including:
  • Plate filling from reagent reservoirs
  • Serial dilutions, plate-to-plate transfers, tube-to-tube transfers, and plate replication
  • Well-to-well transfers (cherry picking protocol for single-channel head only). Unit comes with a single-channel and 12-channel head installed.
  • Accurately and precisely transferring liquids and compounds between microplates
  • Transferring by row or by column
More complex applications can be performed such as sample preparation, timed reaction, PCR preparation, and cell-based assays set up.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 μL < ±2.0%; 100 μL < ±1.5%; 200 μL < ±1.0%
Single-channel, 12-channel, 24-channel and custom-channel heads
1 to 200 μL
Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor, Touch screen controller with software and license, External power supply, Portrait adapter stainless steel plate, Power cords (US, EU, UK, CH), Instruction manual, Calibration certificate
20 μL <1.5% CV; 100 μL <1.0% CV; 200 μL <0.7% CV
100/240 V
Semi-Automated Benchtop Pipettor
1, 12
Corning™ Lambda™ EliteMax pipet tips
96-well standard, shallow, deep well, microcentrifuge, V-, U-, and flat-bottom
5°C to 40°C (relative humidity 80°C% up to 30°C or 45% relative humidity at 40°C)
47 to 63 Hz