Learn More
Corning™ Lambda™ EliteTouch™ 8 Multi-Channel Pipettors, Autoclavable
GREENER_CHOICE
Corning™ Lambda™ EliteTouch™ 8 Multi-Channel Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision.
£549.51 - £570.00
Specifications
Certifications/Compliance ISO8655
Autoclavable Autoclavable
Grip Type Ergonomic
Number of Channels 8
Product Line Lambda EliteTouch
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
16399911
GREENER_CHOICE
View Documents
Corning™
6057
0.5 to 10 μL
£549.51
Each
In Stock
to see stock.
Add to basket
16309921
GREENER_CHOICE
View Documents
Corning™
6058
5-50 μL
£549.51
Each
In Stock
to see stock.
Add to basket
16319921
GREENER_CHOICE
View Documents Promotion Details
Corning™
6059
10 to 200 μL
£560.00
Each
In Stock
to see stock.
Add to basket
16329921
GREENER_CHOICE
View Documents Promotion Details
Corning™
6060
30 to 300 μL
£570.00
Each
In Stock
to see stock.
Add to basket
Description
Description
Corning™ Lambda™ EliteTouch™ pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision. Lightweight construction, contoured shape of the handle, and 4-digit counter were designed to ensure comfortable pipetting. All pipettors feature smooth plunger movement and extremely low pipetting forces to reduce the wrist strain and fatigue (RSI). Colored pushbuttons (included) can help define user or application for sample safety and lower risk of cross contamination.
  • Ergonomic handle design for perfect fit for both right and left hand
  • Convenient one-hand volume setting with auto-lock system prevents accidental volume change during work
  • 4-digit counter for greater precision, perfectly positioned for visibility at all times
  • Single-channel pipettors are compatible with very narrow tubes
  • Fully autoclavable and UV-resistant for sample protection against contaminations
  • The multi-channel shafts retract individually for perfect tip loading and easier tip ejection
  • Easy in-lab calibration helps you quickly adjust your pipettor to various liquid types
  • Individually tested and supplied with a certificate of quality
Learn more from North America
Learn more from Europe, Middle East, and Africa
Specifications
Specifications
ISO8655
Ergonomic
Lambda EliteTouch
Ergonomic
1
Autoclavable
8
Pipette
Instruction manual, QC Certificate, In-lab calibration tool, 3 color pushbuttons
3 Years
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Corning™ Lambda™ EliteTouch™ 8 Multi-Channel Pipettors, Autoclavable