missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B1 Monoclonal Antibody (L-5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 332000
This item is not returnable.
View return policy
Description
Lamin B1 Monoclonal antibody specifically detects Lamin B1 in Chicken, Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Lamin B1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P14731, P14733, P20700, P70615 | |
Lmnb1 | |
Extracts from chicken nuclei | |
100 μg | |
Primary | |
Chicken, Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
L-5 | |
Unconjugated | |
Lmnb1 | |
0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg | |
Mouse | |
Protein A | |
RUO | |
116685, 16906, 396223, 4001 | |
-20°C | |
Liquid |